Hyamine® 1622 0.004 mol/l, Titripur® volumetric solution, for the determination of anionic tensides, Supelco®

Supplier: MilliporeSigma

Synonyms: Hyamine® 1622, N-Benzyl-N,N-dimethyl-2-(2-(4-(2,4,4-trimethylpentan-2-yl)phenoxy)ethoxy)ethanaminium chloride

M-Clarity™ quality level = MQ100

1.15480.1000
EM1.15480.1000CS 880 USD
EM1.15480.1000
Hyamine® 1622 0.004 mol/l, Titripur® volumetric solution, for the determination of anionic tensides, Supelco®
Benzethonium chloride
Formula: C₂₇H₄₂ClNO₂
MW: 448.09 g/mol
Density: 1.0 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011742
CAS Number: 121-54-0
EINECS: 204-479-9

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.0 g/cm3 (20 °C)
pH 9 - 10 (H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR