Water, LiChrosolv® for chromatography, for LC-MS, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ100

1.15333.1000 1.15333.2500
EM1.15333.1000CS 410 USD
EM1.15333.1000 EM1.15333.2500
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Water

LiChrosolv® hypergrade is accurately tested for LC-MS suitability, and meet all the requirements of modern LC-MS ionization methods (ESI/APCI - positive and negative mode).


Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionization efficiency. The packaging material has been improved to meet LC-MS quality requirements perfectly.

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR