Crack Set 10 for the digestion of lead, cadmium, iron, copper, nickel, phosphorus (total) and zinc 100 digestions Spectroquant®
Supplier: MilliporeSigma
1.14687.0001
EM1.14687.0001CS 2197 USD
EM1.14687.0001
Crack Set 10 for the digestion of lead, cadmium, iron, copper, nickel, phosphorus (total) and zinc 100 digestions Spectroquant®
Environmental Test Kits