MColortest™ Sulfate Test, MilliporeSigma
Supplier: MilliporeSigma
1.14411.0001
EM1.14411.0001EA
364
USD
EM1.14411.0001
MColortest™ Sulfate Test, MilliporeSigma
Environmental Test Kits
When the reagents of the MColortestTM test are used up, the kit can be replenished by ordering the appropriate refill pack
Refill packs are available for a range of MColortestTM test kits, making it a particularly economical system.