Iron Test Method: Colorimetric with Color Card and Comparator Block with Long Test Tubes 0.01 - 0.02 - 0.03 - 0.04 - 0.06 - 0.08 - 0.10 - 0.15 -0.20 mg/L Fe MColortest™, MilliporeSigma
Supplier: MilliporeSigma
1.14403.0001
EM1.14403.0001CS 2346 USD
EM1.14403.0001
Iron Test Method: Colorimetric with Color Card and Comparator Block with Long Test Tubes 0.01 - 0.02 - 0.03 - 0.04 - 0.06 - 0.08 - 0.10 - 0.15 -0.20 mg/L Fe MColortest™, MilliporeSigma
Environmental Test Kits