Acetonitrile, LiChrosolv®, Isocratic Grade for liquid chromatography, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

M-Clarity™ quality level = MQ100

1.14291.1000 1.14291.2500 1.14291.4000
EM1.14291.1000CS 1041 USD
EM1.14291.1000 EM1.14291.2500 EM1.14291.5000 EM1.14291.4000
Acetonitrile, LiChrosolv®, Isocratic Grade for liquid chromatography, Supelco®
Acetonitrile

Offering a high degree of UV transmittance, low particle count, low acidity and alkalinity, and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. They are produced from specially selected raw materials, and undergo a number of purification steps prior to final packaging. Since separations are normally carried out under gradient conditions in analytical HPLC, Merck Millipore offer solvents in ’gradient grade’ as well as ’isocratic grade’. This enables you to minimise the gradient effect of the solvent involved.


Offering a high degree of UV transmittance, low particle count, low acidity and alkalinity, and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. They are produced from specially selected raw materials, and undergo a number of purification steps prior to final packaging. Since separations are normally carried out under gradient conditions in analytical HPLC, Merck Millipore offer solvents in ’gradient grade’ as well as ’isocratic grade’. This enables you to minimise the gradient effect of the solvent involved.


The LC-MS LiChrosolv® hypergrade quality solvents are accurately tested for LC-MS suitability, and meet all the requirements of modern LC-MS ionisation methods (ESI/ APCI – positive and negative mode) with low level of ionic background and low ion suppression, they ensure high reproducibility and high ionisation efficiency.

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: -45.7
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR