Deuterium oxide (99.9% D), MagniSolv™ for NMR spectroscopy , pH 7 (H₂O, 20 °C), Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

1.13366.0025 1.13366.0100 1.13366.0500
EM1.13366.0009EA 111 USD
EM1.13366.0009 EM1.13366.0010 EM1.13366.0025 EM1.13366.0100 EM1.13366.0500
Deuterium oxide (99.9% D), MagniSolv™ for NMR spectroscopy , pH 7 (H₂O, 20 °C), Sigma-Aldrich®
Deuterium oxide

MagniSolv™ deuterated solvents provide:
• Reliable results in the NMR-spectra by excellent chemical purity and highest isotopic enrichment.
• Reliable deuteration degrees.
• Low residual water content.
• Determination of water content in two ways.

Formula: D₂O
Boiling Pt: 101.3 °C (1013 hPa)
Melting Pt: 3.8 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9
Merck Index: 12,02984

Order Now

Specification Test Results

Deuteration degree ≥ 99.90 %
Performance test (NMR-spectrum) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR