Lithium metaborate, Spectromelt® A20 for XRF (X-ray fluorescence), fused, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ100

1.12996.1000
EM1.12996.1000CS 25681 USD
EM1.12996.1000
Lithium metaborate, Spectromelt® A20 for XRF (X-ray fluorescence), fused, Supelco®
Lithium metaborate
Formula: BLiO₂
MW: 49.75 g/mol
Boiling Pt: 1719 °C (1013 hPa)
Melting Pt: 845 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011089
CAS Number: 13453-69-5
EINECS: 236-631-5
Merck Index: 13,05546

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 840 °C
Molar Mass 49.75 g/mol
Bulk Density 1300 kg/m3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR