di-Lithium tetraborate, Spectromelt® A 100 for XRF (X-ray fluorescence), Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

1.12630.1000
EM1.12630.1000EA 597 USD
EM1.12630.1000 EM1.12630.5000
di-Lithium tetraborate, Spectromelt® A 100 for XRF (X-ray fluorescence), Supelco®
di-Lithium tetraborate

Spectromelt is based on lithium borates for fluxing digestion as granulate or as fused material used for X-ray fluorescence analysis (XRF).
The fused particles are spherical, have a small particle size distribution and a higher bulk density.
The material is easy to handle and has a low loss on ignition.


  • Extensive specification of trace impurities
  • Batch-to-batch consistency
  • High purity materials

Formula: Li₂B₄O₇
Melting Pt: 918 °C
Density: 1.4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011083
CAS Number: 12007-60-2
EINECS: 234-514-3
Merck Index: 14,05524

Order Now

Specification Test Results

Assay (acidimetric) ≥ 99.0 %
Chloride (Cl) ≤ 50 ppm
Phosphate (PO₄) ≤ 20 ppm
Silicate (SiO₂) ≤ 100 ppm
Sulphate (SO₄) ≤ 50 ppm
Ag (Silver) ≤ 5 ppm
Al (Aluminium) ≤ 10 ppm
Ba (Barium) ≤ 5 ppm
Ca (Calcium) ≤ 20 ppm
Cd (Cadmium) ≤ 5 ppm
Co (Cobalt) ≤ 5 ppm
Cr (Chromium) ≤ 5 ppm
Cu (Copper) ≤ 5 ppm
Fe (Iron) ≤ 10 ppm
Ga (Gallium) ≤ 5 ppm
K (Potassium) ≤ 10 ppm
Mg (Magnesium) ≤ 5 ppm
Mn (Manganese) ≤ 5 ppm
Na (Sodium) ≤ 20 ppm
Ni (Nickel) ≤ 5 ppm
Pb (Lead) ≤ 5 ppm
Sr (Strontium) ≤ 5 ppm
Zn (Zinc) ≤ 5 ppm
Loss on ignition (1000 °C) ≤ 0.2 %
Bulk density about 120


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR