Holmium(III) oxide, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Diholmium trioxide, Holmium oxide

M-Clarity™ quality level = MQ200

1.12469.0002
EM1.12469.0002EA 115 USD
EM1.12469.0002
Holmium(III) oxide, Sigma-Aldrich®
Holmium (III) oxide
Formula: Ho₂O₃
Boiling Pt: 2600 °C (1013 hPa)
Melting Pt: 2400 °C
Density: 3.79 g/cm³ (20 °C)
MDL Number: MFCD00011053
CAS Number: 12055-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR