Tellurium dioxide ≥99.5%, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
1.12356.0025
1.12356.1000
EM1.12356.0025CS
6831
USD
EM1.12356.0025
EM1.12356.1000
Tellurium dioxide ≥99.5%, Sigma-Aldrich®
Tellurium dioxide
Formula:
O₂Te |
MDL Number:
MFCD00011263 CAS Number: 7446-07-3 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 733 °C |
Molar Mass | 159.60 g/mol |
Boiling Point | 1245 °C |
Density | 6.02 g/cm3 (20 °C) |
Synonyms | Tellurium(IV) oxide |
Hill Formula | O2Te |
Chemical Formula | TeO2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...