Neodymium(III) oxide ≥99%, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide

M-Clarity™ quality level = MQ200

1.12276.0025
EM1.12276.0025CS 3729 USD
EM1.12276.0025
Neodymium(III) oxide ≥99%, Sigma-Aldrich®
Neodymium (III) oxide
Formula: Nd₂O₃
Boiling Pt: 3760 °C (1013 hPa)
Melting Pt: 2270 °C
Density: 7.24 g/cm³ (20 °C)
MDL Number: MFCD00011134
CAS Number: 1313-97-9
EINECS: 215-214-1
Merck Index: 14,06450

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 2272 °C
Molar Mass 336.48 g/mol
Bulk Density 1200 kg/m3
Density 7.24 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR