Sodium octyl sulfate, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Octyl sulphate, sodium salt, Octyl sulfate, sodium salt, Octyl sulfuric acid salt, Octyl sulphuric acid salt

M-Clarity™ quality level = MQ200

1.12268.0500
EM1.12268.0500EA 14924 USD
EM1.12268.0500
Sodium octyl sulfate, Sigma-Aldrich®
Sodium octyl sulphate
Formula: C₈H₁₇NaO₄S
Melting Pt: 181…183 °C
Density: 1.266 g/cm³ (20 °C)
MDL Number: MFCD00007470
CAS Number: 142-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR