Sodium octyl sulfate

Supplier: MilliporeSigma

Synonyms: Octyl sulphate, sodium salt, Octyl sulfate, sodium salt, Octyl sulfuric acid salt, Octyl sulphuric acid salt

1.12268.0005
EM1.12268.0005CS 6849 USD
EM1.12268.0005
Sodium octyl sulfate
Sodium octyl sulphate

Octyl sulfate sodium salt for surfactant tests.

Formula: C₈H₁₇NaO₄S
MDL Number: MFCD00007470
CAS Number: 142-31-4

Order Now

Specification Test Results

Hill Formula C₈H₁₇NaO₄S
Chemical formula C₈H₁₇NaSO₄
Molar Mass 232.26 g/mol
Melting Point 195 °C (decomposition)
pH value 6 - 8 (10 g/l, H₂O, 20 °C)
Bulk density 470 kg/m³
Solubility (20 °C) soluble
LD 50 oral LD50 Rat 3200 mg/kg
Storage class 10 - 13 Other liquids and solids
WGK WGK 2 water endangering


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR