Molybdenum ≥99%, powder, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

1.12254.0100
EM1.12254.0100CS 25041 USD
EM1.12254.0100
Molybdenum ≥99%, powder, Sigma-Aldrich®
Molybdenum

Molybdenum powder 99+

Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
MDL Number: MFCD00003465
CAS Number: 7439-98-7
UN: 3089
Merck Index: 12,06317

Order Now

Specification Test Results

Hill Formula Mo
Molar Mass 95.94 g/mol
Density 10.2 g/cm³ (20 °C)
Melting Point 2620 °C
Bulk density 1400 kg/m³
Solubility (20 °C) insoluble
Storage class 10 - 13 Other liquids and solids
WGK NWG not water endangering


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR