Manganese ≥99%, powder, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

1.12237.0250
EM1.12237.0250CS 9179 USD
EM1.12237.0250
Manganese ≥99%, powder, Sigma-Aldrich®
Manganese
Formula: Mn
MDL Number: MFCD00011111
CAS Number: 7439-96-5
UN: 3089
ADR: 4.1,

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 1245 °C
Molar Mass 54.94 g/mol
Boiling Point 2097 °C
Density 7.2 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR