Lanthanum(III) oxide, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

1.12220.0100 1.12220.0500
EM1.12220.0100CS 23387 USD
EM1.12220.0100 EM1.12220.0500
Lanthanum(III) oxide, Supelco®
Lanthanum (III) oxide

EMD # 1.12220.0500 replaces and is identical to EMD # LX0045-2.

Formula: La₂O₃
MDL Number: MFCD00011071
CAS Number: 1312-81-8

Order Now

Specification Test Results

Melting Point 2315 °C
Molar Mass 325.82 g/mol
Bulk Density 1300 kg/m3
Boiling Point 4200 °C
Density 6.51 g/cm3 (20 °C)
pH 9.0 (50 g/l, H2O, 20 °C) (slurry)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR