Indium (III) oxide ≥99.5%, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Indium trioxide, Diindium trioxide

M-Clarity™ quality level = MQ200

1.12201.0010
EM1.12201.0010CS 6454 USD
EM1.12201.0010
Indium (III) oxide ≥99.5%, Sigma-Aldrich®
Indium (III) oxide
Formula: In₂O₃
MDL Number: MFCD00011060
CAS Number: 1312-43-2

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 2000 °C
Molar Mass 277.64 g/mol
Density 7.18 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR