Indium (III) oxide ≥99.5%, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Indium trioxide, Diindium trioxide

M-Clarity™ quality level = MQ200

1.12201.0005
EM1.12201.0005EA 91 USD
EM1.12201.0005
Indium (III) oxide ≥99.5%, Sigma-Aldrich®
Indium (III) oxide
Formula: In₂O₃
Melting Pt: 715.05 °C
Density: 7.18 g/cm³ (20 °C)
MDL Number: MFCD00011060
CAS Number: 1312-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR