Dysprosium(III) oxide ≥99%, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Didysprosium trioxide, Dysprosium oxide

M-Clarity™ quality level = MQ200

1.12151.0010
EM1.12151.0010CS 49954 USD
EM1.12151.0010
Dysprosium(III) oxide ≥99%, Sigma-Aldrich®
Dysprosium (III) oxide
Formula: Dy₂O₃
MW: 373 g/mol
Melting Pt: 2400
Density: 7.81 g/cm³ (20 °C)
MDL Number: MFCD00010986
CAS Number: 1308-87-8
EINECS: 215-164-0
Merck Index: 14,03482

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 2340 °C
Molar Mass 372.99 g/mol
Density 7.81 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR