Potassium hydroxide 2 mol/l (2 N) in methanol, Titripur® volumetric solution, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

M-Clarity™ quality level = MQ100

1.11787.2500
EM1.11787.2500CS 502 USD
EM1.11787.2500
Potassium hydroxide 2 mol/l (2 N) in methanol, Titripur® volumetric solution, Supelco®
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 3286
ADR: 3 (6.1, 8),II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR