Formic acid 98-100%, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Aminic Acid, Methanoic acid

M-Clarity™ quality level = MQ100

1.11670.1000
EM1.11670.1000CS 4175 USD
EM1.11670.1000 EM1.11670.0250
Formic acid 98-100%, Suprapur® for trace metal analysis, Supelco®
Formic acid
Formula: HCO₂H
MW: 46.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8 (3),
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR