N,N-Dimethylformamide-[D₇] ≥99.5% (Isotopic) 99.5, MagniSolv™ for NMR spectroscopy for NMR spectroscopy, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Heptadeutero-N,N-dimethylformamide, N,N-Di[2H3]methyl[2H]formamide, N,N-Dimethylformamide-d7, DMF-D7, DMF-d7, N,N-Dimethylformamide-[²H₇], Formdimethylamide-D7, Formic acid dimethylamide-D7

M-Clarity™ quality level = MQ200

1.11656.0001 1.11656.0009
EM1.11656.0001EA 197 USD
EM1.11656.0001 EM1.11656.0009
N,N-Dimethylformamide-[D₇] ≥99.5% (Isotopic) 99.5, MagniSolv™ for NMR spectroscopy for NMR spectroscopy, Sigma-Aldrich®
N,N-Dimethylformamide-[D7]

MagniSolv™ deuterated solvents provide reliable results in the NMR-spectra with extremely low residual water, excellent chemical purity and the highest isotopic enrichment.

Formula: DCOON(CD₃)₂
Boiling Pt: 152 °C (1013 hPa)
Melting Pt: –60 °C
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00003286
CAS Number: 4472-41-7
EINECS: 224-745-8
UN: 2265
ADR: 3,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR