Sodium hydroxide 45% in aqueous solution, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

M-Clarity™ quality level = MQ300

1.11360.2500 1.11360.9025
EM1.11360.2500CS 638 USD
EM1.11360.2500 EM1.11360.9025
Sodium hydroxide 45% in aqueous solution, EMSURE® for analysis, Supelco®
Sodium hydroxide
Formula: NaOH
Boiling Pt: 135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.48 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 7 °C
Boiling Point 135 °C (1013 hPa)
Density 1.48 g/cm3 (20 °C)
pH 14 (89 g/l, H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR