Sodium hydroxide 45% in aqueous solution, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
M-Clarity™ quality level = MQ300
1.11360.2500
1.11360.9025
EM1.11360.2500CS
638
USD
EM1.11360.2500
EM1.11360.9025
Sodium hydroxide 45% in aqueous solution, EMSURE® for analysis, Supelco®
Sodium hydroxide
Formula:
NaOH Boiling Pt: 135 °C (1013 hPa) Melting Pt: 9…10 °C Density: 1.48 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 7 °C |
Boiling Point | 135 °C (1013 hPa) |
Density | 1.48 g/cm3 (20 °C) |
pH | 14 (89 g/l, H2O, 20 °C) |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...