MColortest™ Chloride Tests, MilliporeSigma
Supplier: MilliporeSigma
1.11106.0001
EM1.11106.0001EA 198 USD
EM1.11106.0001
MColortest™ Chloride Tests, MilliporeSigma
Environmental Test Kits
Chloride is found in the earth's crust and can therefore also be found in all natural waters at some concentration
  • When the reagents of the MColortest™ test are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of MColortest™ test kits, making it a particularly economical system.
This basic parameter of water analysis can be checked using the Chloride Test.