Carbonate Hardness Test Method: Colorimetric With Test Strips 4 - 8 - 12 - 16 - 24°d MQuant™, MilliporeSigma
Supplier: MilliporeSigma
1.10648.0001
EM1.10648.0001CS 22657 USD
EM1.10648.0001
Carbonate Hardness Test Method: Colorimetric With Test Strips 4 - 8 - 12 - 16 - 24°d MQuant™, MilliporeSigma
Environmental Test Kits
The test is suitable for rapid determination of carbonate hardness
MilliporeSigma # 1.10648.0001 replaces and is identical to MilliporeSigma # 10648-1
Carbonate hardness is part of the total hardness of water and is defined as that portion of alkaline earth ions present in the water for which there exists an equivalent amount of hydrogen carbonate ions and carbonate ions originating from dissolved carbonic acid.