Ascorbic Acid Test Method: Colorimetric With Test Strips 50 - 100 - 200 - 300 - 500 - 700 - 1000 - 2000 mg/L MQuant™, MilliporeSigma
Supplier: MilliporeSigma
1.10023.0001
EM1.10023.0001CS 12066.13 USD
EM1.10023.0001
Ascorbic Acid Test Method: Colorimetric With Test Strips 50 - 100 - 200 - 300 - 500 - 700 - 1000 - 2000 mg/L MQuant™, MilliporeSigma
Environmental Test Kits
The ascorbic acid test is suitable for quickly measuring levels of natural ascorbic acid (vitamin C) in foods such as fruit and vegetable juices, soft drinks, and also to check how much ascorbic acid has been added to certain foods as a preservative or antioxidant.
MilliporeSigma # 1.10023.0001 replaces and is identical to MilliporeSigma # 10023-1