Sulfite Test Method: Colorimetric With Test Strips 10 - 40 - 80 - 180 - 400 mg/L SO32- MQuant™, MilliporeSigma
Supplier: MilliporeSigma
1.10013.0001 1.10013.0001
CA1.10013.0001EA 176.54 USD
CA1.10013.0001 EM1.10013.0001
Sulfite Test Method: Colorimetric With Test Strips 10 - 40 - 80 - 180 - 400 mg/L SO32- MQuant™, MilliporeSigma
Environmental Test Kits
The sulfite test is suitable for food analysis, particularly in the testing of wine and fruit juices, sulfurated foods (e.g
MilliporeSigma # 1.10013.0001 replaces and is identical to MilliporeSigma # 10013-1
fresh and dried fruits), fish shellfish, crustaceans and minced meat products.