Cobalt Test Method: Colorimetric With Test Strips 10 - 30 - 100 - 300 - 1000 mg/L Co²⁺ MQuant™, MilliporeSigma
Supplier: MilliporeSigma
1.10002.0001
EM1.10002.0001CS 26266 USD
EM1.10002.0001
Cobalt Test Method: Colorimetric With Test Strips 10 - 30 - 100 - 300 - 1000 mg/L Co²⁺ MQuant™, MilliporeSigma
Environmental Test Kits
The cobalt test can be used in the testing of waste water, electroplating baths, in the electronics industry, metal industry, in the assaying and refining of ores.
MilliporeSigma # 1.10002.0001 replaces and is identical to MilliporeSigma # 10002-1