Silver nitrate 0.1 M concentrated aqueous solution, Titrisol®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

1.09990.0001
EM1.09990.0001CS 1749 USD
EM1.09990.0001
Silver nitrate 0.1 M concentrated aqueous solution, Titrisol®, Supelco®
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1760
ADR: 8,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.27 g/cm3 (20 °C)
pH 7 - 9 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR