Aluminium Standard, Titrisol®, MilliporeSigma
Supplier: MilliporeSigma
1.09967.0001
EM1.09967.0001CS 4628 USD
EM1.09967.0001
Aluminium Standard, Titrisol®, MilliporeSigma
Standards Cationic Standards Aluminum Standards
1000 mg of aluminium (as AlCl₃ in H₂O) in a plastic ampule for dilution.
pH 2.5 (H₂O, 20 °C)