Barium chloride 0.1 N concentrated aqueous solution, Titrisol®, Supelco®

Supplier: MilliporeSigma
Warning

1.09962.0001
EM1.09962.0001CS 2391 USD
EM1.09962.0001
Barium chloride 0.1 N concentrated aqueous solution, Titrisol®, Supelco®
Barium chloride
Formula: BaCl₂
MW: 208.24 g/mol
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
UN: 3287
ADR: 6.1,

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.18 g/cm3 (20 °C)
pH 4.3 (H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR