Sodium thiosulfate 0.1 M aqueous solution_concentrated, Titrisol®, Supelco®

Supplier: MilliporeSigma

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

1.09950.0001
EM1.09950.0001CS 475 USD
EM1.09950.0001
Sodium thiosulfate 0.1 M aqueous solution_concentrated, Titrisol®, Supelco®
Sodium thiosulphate
Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.22 g/cm3 (20 °C)
pH 9 - 10 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR