Sodium chloride 0.1 M concentrated aqueous solution, Titrisol®, Supelco®

Supplier: MilliporeSigma

1.09945.0001
EM1.09945.0001CS 1665 USD
EM1.09945.0001
Sodium chloride 0.1 M concentrated aqueous solution, Titrisol®, Supelco®
Sodium chloride
Formula: NaCl
Density: 1.08 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now

Specification Test Results

Concentration after dilution to 1 liter - C (NaCl) 0.1 mol/l
Amount of substance concentration 0.1000 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR