Calcium Standard, Titrisol®, MilliporeSigma
Supplier: MilliporeSigma
1.09943.0001
EM1.09943.0001CS
4502
USD
EM1.09943.0001
Calcium Standard, Titrisol®, MilliporeSigma
Standards
Cationic Standards
Calcium Standards
CaCl₂ in 6,5% hydrochloric acid
Traceable to SRM from NIST