Oxalic acid concentrate for dilution to 0.005 mol/L (0.01 N), Titrisol®, Supelco®

Supplier: MilliporeSigma

Synonyms: Ethanedioic acid

Dose for preparation of 1 litre of solution.

1.09932.0001
EM1.09932.0001CS 1755 USD
EM1.09932.0001
Oxalic acid concentrate for dilution to 0.005 mol/L (0.01 N), Titrisol®, Supelco®
Oxalic acid
Formula: C₂H₂O₄
MW: 90.04 g/mol
Boiling Pt: 365
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
Merck Index: 13,06980

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.002 g/cm3 (20 °C)
pH 1.5 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR