Potassium hydroxide 0.1 M in aqueous solution, Titrisol®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

1.09921.0001
EM1.09921.0001CS 1860 USD
EM1.09921.0001
Potassium hydroxide 0.1 M in aqueous solution, Titrisol®, Supelco®
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.09 g/cm3 (20 °C)
pH 14 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR