Sodium hydroxide 5 M concentrated aqueous solution, Titrisol®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

M-Clarity™ quality level = MQ100

1.09913.0001
EM1.09913.0001CS 378 USD
EM1.09913.0001
Sodium hydroxide 5 M concentrated aqueous solution, Titrisol®, Supelco®
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 109 °C (1013 hPa)
Density: ≤1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.185 g/cm3 (20 °C)
pH 14 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR