Ammonium thiocyanate 0.1 M concentrated aqueous solution, Titrisol®, Supelco®

Supplier: MilliporeSigma

Synonyms: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate, Ammonium sulfocyanate

1.09900.0001
EM1.09900.0001CS 2081 USD
EM1.09900.0001
Ammonium thiocyanate 0.1 M concentrated aqueous solution, Titrisol®, Supelco®
Ammonium thiocyanate
Formula: NH₄SCN
MW: 76.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS Number: 1762-95-4
EINECS: 217-175-6

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.03 g/cm3 (20 °C)
pH 4.5 (H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR