Titriplex® solution B for the determination of alkaline earth in water (hardness) concentrate, Titrisol®, Supelco®
Supplier: MilliporeSigma
Warning
1.09895.0001
EM1.09895.0001CS 2422 USD
EM1.09895.0001
Titriplex® solution B for the determination of alkaline earth in water (hardness) concentrate, Titrisol®, Supelco®
Titriplex® solution B
for the determination of alkaline earth in water (hardness), 1 ml = 10 mg CaO/l using 100 ml of water for 1000 ml