Eosin Y (yellowish) 0.5% in aqueous solution for microscopy, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Solvent Red 43, Field's Stain B, Eosin 225, 2,4,5,7-Tetrabromofluorescein disodium salt, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Tetrabromofluorescein disodium salt, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Eosin Y disodium salt

1.09844.2500
EM1.09844.1000CS 665.56 USD
EM1.09844.1000 EM1.09844.2500
Eosin Y (yellowish) 0.5% in aqueous solution for microscopy, Sigma-Aldrich®
Eosin Y (yellowish)
Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
MDL Number: MFCD00005040
CAS Number: 17372-87-1

Order Now

Specification Test Results

Density 1.00 g/cm3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR