Methyl oleate reference substance for gas chromatography, Supelco®

Supplier: MilliporeSigma

Synonyms: cis-Oleic Acid Methyl Ester, Oleic acid methyl ester

1.09743.0005
EM1.09743.0005CS 83516 USD
EM1.09743.0005
Methyl oleate reference substance for gas chromatography, Supelco®
Methyl oleate
Formula: C₁₉H₃₆O₂
MW: 296.49 g/mol
MDL Number: MFCD00009578
CAS Number: 112-62-9

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -20 °C
Molar Mass 296.49 g/mol
Boiling Point 202 - 210 °C (20 hPa)
Flash Point 113 °C
Density 0.87 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR