n-Pentane reference substance for gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Pentane

1.09719.0005
EM1.09719.0005CS 84373 USD
EM1.09719.0005
n-Pentane reference substance for gas chromatography, Supelco®
n-Pentane
Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 35…36 °C (1013 hPa)
Melting Pt: –130 °C
Density: 0.63 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.04 g/l (25 °C)
Melting Point -130 °C
Molar Mass 72.15 g/mol
Boiling Point 36 °C (1013 hPa)
Vapor Pressure 570 hPa (20 °C)
Flash Point -48 °C (refers to pure substance)
Refractive Index 1.358
Explosion Limit 1.4 - 8 %(V)
Density 0.63 g/cm3 (20 °C)
pH (H2O) no data available
Ignition Point 285 °C
Hazard Class 3
Hazard Class Description Flammable liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR