Methyl ethyl ketone reference substance for gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: EK 500 Additive, Butan-2-one, Ethyl methyl ketone, 2-Butanone, Butanone, MEK

1.09709.0005
EM1.09709.0005CS 65903 USD
EM1.09709.0005
Methyl ethyl ketone reference substance for gas chromatography, Supelco®
Methyl ethyl ketone
Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 79.6 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0.8054 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3,
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 292 g/l (20 °C)
Melting Point -86 °C
Molar Mass 72.11 g/mol
Boiling Point 79.6 °C (1013 hPa)
Vapor Pressure 105 hPa (20 °C)
Flash Point -1 °C
Refractive Index 1.3814 (15 °C)
Explosion Limit 1.8 - 11.5 %(V)
Density 0.805 g/cm3 (20 °C)
pH 5.5 (300 g/l, H2O, 20 °C)
Ignition Point 514 °C
Saturation concentration 310 g/m3 (20 °C) Air
Evaporation number 6.3
Hazard Class 3
Hazard Class Description Flammable liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR