n-Hexane reference substance for gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger

1.09687.0005
EM1.09687.0005CS 84157 USD
EM1.09687.0005
n-Hexane reference substance for gas chromatography, Supelco®
n-Hexane
Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –94.3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.0095 g/l (20 °C)
Melting Point -94.3 °C
Molar Mass 86.18 g/mol
Boiling Point 69 °C (1013 hPa)
Vapor Pressure 160 hPa (20 °C)
Flash Point -22 °C
Refractive Index 1.375
Explosion Limit 1.0 - 8.1 %(V)
Density 0.66 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 240 °C
Viscosity kinematic 0.50 mm2/s (20 °C)
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR