2',7'-Dichlorofluorescein indicator, Supelco®

Supplier: MilliporeSigma

1.09676.0005
EM1.09676.0005CS 51369 USD
EM1.09676.0005
2',7'-Dichlorofluorescein indicator, Supelco®
2',7'-Dichlorofluorescein
Formula: C₂₀H₁₀Cl₂O₅
MDL Number: MFCD00005047
CAS Number: 76-54-0
EINECS: 200-968-6

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 401.2 g/mol
Bulk Density 390 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR