p-Toluenesulfonic acid monohydrate, EMSURE® ACS for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: 4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate, 4-Methylbenzenesulfonic acid hydrate, Toluene-4-sulfonic acid monohydrate

1.09613.0100 1.09613.0500
EM1.09613.0100CS 3253 USD
EM1.09613.0100 EM1.09613.0500
p-Toluenesulfonic acid monohydrate, EMSURE® ACS for analysis, Supelco®
p-Toluenesulphonic acid monohydrate

In North America 1.09613.0500 replaces and is identical to TX0770-3

Formula: C₇H₈O₃S·1H₂O
Melting Pt: 102…104 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00142137
CAS Number: 6192-52-5
EINECS: 203-180-0
UN: 2585
ADR: 8,
Merck Index: 12,09671

Order Now

Specification Test Results

Solubility 670 g/l (20 °C)
Melting Point 56 °C Elimination of water of crystallisation
Molar Mass 190.22 g/mol
Bulk Density 510 kg/m3
Boiling Point 140 °C (27 hPa)
Flash Point 180 °C
pH 1 (650 g/l, H2O, 20 °C) (anhydrous substance)
Ignition Point 600 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR