n-Tetradecane reference substance for gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger

1.09608.0005
EM1.09608.0005CS 79521 USD
EM1.09608.0005
n-Tetradecane reference substance for gas chromatography, Supelco®
n-Tetradecane
Formula: H₃C(CH₂)₁₂CH₃
Boiling Pt: 253.7 °C (1013 hPa)
Melting Pt: 6 °C
Density: 0.7628 g/cm³ (20 °C)
Flash Pt: 102 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008986
CAS Number: 629-59-4
EINECS: 211-096-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03064a29015d1e810f1e4350a7a6cd2dfe4713fd&vid=f4838a5686fe033622495a3bf280963555654d9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility  - 0.0001 g/l (20 °C)
Melting Point 6 °C
Molar Mass 198.39 g/mol
Boiling Point 254 °C
Vapor Pressure 1.55 Pa (25 °C)
Flash Point 102 °C
Explosion Limit 0.5 - 6.5 %(V)
Density 0.76 g/cm3 (20 °C)
Ignition Point 202 °C
Viscosity kinematic 3 mm2/s (40 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR