n-Decane reference substance for gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger

1.09603.0005
EM1.09603.0005CS 81858 USD
EM1.09603.0005
n-Decane reference substance for gas chromatography, Supelco®
n-Decane
Formula: H₃C(CH₂)₈CH₃
Boiling Pt: 174.1 °C (1013 hPa)
Melting Pt: –30 °C
Density: 0.73 g/cm³ (20 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00008954
CAS Number: 124-18-5
EINECS: 204-686-4
UN: 2247
ADR: 3,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e4dead3a7841a2258e8a5b99b880895cd06aa1&vid=209e0ebbe6a38426575fcf5608cf6f2b586e6d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.00005 g/l (25 °C)
Melting Point -30 °C
Molar Mass 142.28 g/mol
Boiling Point 174 °C
Vapor Pressure 1 hPa (16 °C)
Flash Point 51 °C
Explosion Limit 0.7 - 5.4 %(V)
Density 0.73 g/cm3 (20 °C)
Ignition Point 208 °C
Saturation concentration 11 g/m3 (20 °C) Air
Viscosity kinematic  - 7 mm2/s (40 °C)
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR